Learn More
ATG4C, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ATG4C.
Brand: Abnova PAB0737.100uL
Description
Autophagy is the process by which endogenous proteins and damaged organelles are destroyed intracellularly. Autophagy is postulated to be essential for cell homeostasis and cell remodeling during differentiation, metamorphosis, nonapoptotic cell death, and aging. Reduced levels of autophagy have been described in some malignant tumors, and a role for autophagy in controlling the unregulated cell growth linked to cancer has been proposed. This gene encodes a member of the autophagin protein family. The encoded protein is also designated as a member of the C-54 family of cysteine proteases. Alternate transcriptional splice variants, encoding the same protein, have been characterized. [provided by RefSeq
Specifications
| ATG4C | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of ATG4C. | |
| Lyophilized from PBS | |
| Q96DT6;Q811C2;109475174 | |
| ATG4C | |
| A synthetic peptide (conjugated with Blue Carrier Protein) corresponding to C-terminus of human ATG4C. | |
| RUO | |
| 84938 | |
| Store at 4°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| Immunofluorescence, Immunohistochemistry (Frozen), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:100-1:2000) Immunofluorescence (1:100-1:2000) Western Blot (1:100-1:2000) The optimal working dilution should be determined by the end user. | |
| ATG4C | |
| APG4-C/APG4C/AUTL1/AUTL3/FLJ14867 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only