missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATG2B Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ATG2B.
Brand: Abnova PAB25638.100ug
This item is not returnable.
View return policy
Specifications
| ATG2B | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| ATG2B | |
| C14orf103/FLJ10242 | |
| Rabbit | |
| Peptide affinity purification | |
| RUO | |
| 55102 | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of ATG2B. | |
| In PBS (0.05% sodium azide) | |
| NP_060506 | |
| ATG2B | |
| A synthetic peptide corresponding to 19 amino acids at N-terminus of human ATG2B. | |
| 100 μg | |
| Primary | |
| ATG2B antibody is predicted to not cross-react with other ATG2A. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction