Learn More
ASGR1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant ASGR1.
Brand: Abnova PAB20573.100uL
Description
Partially deglycosylated plasma glycoproteins and immunoglobulin IgA2 allotypes are efficiently and specifically removed from circulation by a receptor-mediated process. The asialoglycoprotein receptor binds to desialylated (galactosyl-terminal) glycoproteins. It transports these glycoproteins via a series of membrane vesicles and tubules to an acidic-sorting organelle where the receptor and ligand dissociate. Then the receptor is recycled back to the cell surface and the ligand is transported to the lysosomes for degradation. [provided by RefSeq
Sequence: MTKEYQDLQHLDNEESDHHQLRKGPPPPQPLLQRLCSGPRLSpecifications
| ASGR1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant ASGR1. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| P07306 | |
| ASGR1 | |
| Recombinant protein corresponding to amino acids of human ASGR1. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:200-1:500) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| ASGR1 | |
| HL-1/ASGPR/ASGPR1/CLEC4H1 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 432 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only