Learn More
ARID3B, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ARID3B.
Brand: Abnova PAB3144.400uL
Description
This gene encodes a member of the ARID (AT-rich interaction domain) family of DNA-binding proteins. The encoded protein is homologous with two proteins that bind to the retinoblastoma gene product, and also with the mouse Bright and Drosophila dead ringer proteins. A pseudogene on chromosome 1p31 exists for this gene. Members of the ARID family have roles in embryonic patterning, cell lineage gene regulation, cell cycle control, transcriptional regulation and possibly in chromatin structure modification. [provided by RefSeq
Specifications
| ARID3B | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of ARID3B. | |
| In PBS (0.09% sodium azide) | |
| NP_006456;Q8IVW6 | |
| ARID3B | |
| A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human ARID3B. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Western Blot (1:1000) Immunohistochemistry (1:10-50) The optimal working dilution should be determined by the end user. | |
| ARID3B | |
| BDP/DRIL2 | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| 10620 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only