Learn More
ARHGAP31, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant ARHGAP31.
Brand: Abnova PAB22993.100uL
Description
This gene encodes a GTPase-activating protein (GAP). A variety of cellular processes are regulated by Rho GTPases which cycle between an inactive form bound to GDP and an active form bound to GTP. This cycling between inactive and active forms is regulated by guanine nucleotide exchange factors and GAPs. The encoded protein is a GAP shown to regulate two GTPases involved in protein trafficking and cell growth. [provided by RefSeq
Sequence: QDLDIVAHALTGRRNSAPVSVSAVRTSFMVKMCQARAVPVIPPKIQYTQIPQPLPSQSSGENGVQPLERSQEGPSSTSGTTQKPAKDDSPSSLESSKEESpecifications
| ARHGAP31 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant ARHGAP31. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| Q2M1Z3 | |
| ARHGAP31 | |
| Recombinant protein corresponding to amino acids of human ARHGAP31. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:200-1:500) The optimal working dilution should be determined by the end user. | |
| ARHGAP31 | |
| KIAA1204/MGC138368/MGC138370 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 57514 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only