missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARHGAP26, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ARHGAP26.
Brand: Abnova PAB3298.400uL
This item is not returnable.
View return policy
Specifications
| ARHGAP26 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of ARHGAP26. | |
| In PBS (0.09% sodium azide) | |
| Q9UNA1 | |
| ARHGAP26 | |
| A synthetic peptide (conjugated with KLH) corresponding to internal region of human ARHGAP26. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| ARHGAP26 | |
| FLJ42530/GRAF/KIAA0621/OPHN1L/OPHN1L1 | |
| Rabbit | |
| Protein G purification | |
| RUO | |
| 23092 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction