Learn More
ARHGAP23, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant ARHGAP23.
Brand: Abnova PAB21244.100uL
Description
The RHO (see ARHA; MIM 165390) family of small GTPases are involved in signal transduction through transmembrane receptors, and they are inactive in the GDP-bound form and active in the GTP-bound form. GTPase-activating proteins, such as ARHGAP23, inactivate RHO family proteins by stimulating their hydrolysis of GTP (Katoh and Katoh, 2004 [PubMed 15254754]).[supplied by OMIM
Sequence: MNLGFGDESPEPEASGRGERLGRKVAPLATTEDSLASIPFIDEPTSPSIDLQAKHVPASAVVSSAMNSAPVLGTSPSSSpecifications
| ARHGAP23 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant ARHGAP23. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| Q9P227 | |
| ARHGAP23 | |
| Recombinant protein corresponding to amino acids of human ARHGAP23. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| ARHGAP23 | |
| KIAA1501 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 57636 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only