Learn More
ARHGAP12, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of human ARHGAP12.
Brand: Abnova PAB1136.100uL
Description
RHO GTPases (see ARHA, MIM 165390) that participate in a wide variety of cellular processes are active in their GTP-bound form and inactive in their GDP-bound form. The hydrolysis of GTP and the inactivation of these GTPases are regulated by GTPase-activating proteins (GAPs), such as ARHGAP12 (Zhang et al., 2002 [PubMed 11854031]).[supplied by OMIM
Sequence: ILVKKTNDDWWQVKPDSpecifications
| ARHGAP12 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of human ARHGAP12. | |
| In serum (50% glycerol, 0.025% sodium azide) | |
| NP_060757 | |
| ARHGAP12 | |
| A synthetic peptide (conjugated with KLH) corresponding to amino acids 40-56 of human ARHGAP12. | |
| RUO | |
| 94134 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:100-1:1000) Western Blot (2.0 ug/mL) The optimal dilution should be determined by the end user. | |
| ARHGAP12 | |
| DKFZp779N2050/FLJ10971/FLJ20737/FLJ21785/FLJ45709 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only