Learn More
ARFGAP1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ARFGAP1.
Brand: Abnova PAB4255.400uL
Description
The protein encoded by this gene is a GTPase-activating protein (GAP) which associates with the Golgi apparatus and which interacts with ADP-ribosylation factor 1 (ARF1). The encoded protein promotes hydrolysis of ARF1-bound GTP and is required for the dissociation of coat proteins from Golgi-derived membranes and vesicles. Dissociation of the coat proteins is required for the fusion of these vesicles with target compartments. The activity of this protein is stimulated by phosphoinosides and inhibited by phosphatidylcholine. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Specifications
| ARFGAP1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of ARFGAP1. | |
| In PBS (0.09% sodium azide) | |
| NP_060679;Q8N6T3 | |
| ARFGAP1 | |
| A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human ARFGAP1. | |
| 400 μL | |
| Primary | |
| Human, Rat | |
| Liquid |
| ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| ELISA (1:1000) Western Blot (1:100-500) Immunohistochemistry (1:50-100) The optimal working dilution should be determined by the end user. | |
| ARFGAP1 | |
| ARF1GAP/HRIHFB2281/MGC39924 | |
| Rabbit | |
| Protein G purification | |
| RUO | |
| 55738 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only