missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARC, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ARC.
Brand: Abnova PAB13559.100ug
This item is not returnable.
View return policy
Specifications
| ARC | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of ARC. | |
| In PBS (0.02% sodium azide) | |
| Arg3.1/KIAA0278 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| This antibody binds ARC. | |
| Store at 4°C. |
| Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| ARC | |
| ARC | |
| A synthetic peptide corresponding to amino acids 191-208 of human ARC. | |
| RUO | |
| 23237 | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction