Learn More
ARAP2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant ARAP2.
Brand: Abnova PAB22927.100uL
Description
The protein encoded by this gene contains ARF-GAP, RHO-GAP, ankyrin repeat, RAS-associating, and pleckstrin homology domains. The protein is a phosphatidylinositol (3,4,5)-trisphosphate-dependent Arf6 GAP that binds RhoA-GTP, but it lacks the predicted catalytic arginine in the RHO-GAP domain and does not have RHO-GAP activity. The protein associates with focal adhesions and functions downstream of RhoA to regulate focal adhesion dynamics. [provided by RefSeq
Sequence: HCLEHKDDKLRNRPRKHRSFNCLEDTEPEAPLGQPKGHKGLKTLRKTEDRNSKATLDSDHKLPSRVIEELNVVLQRSRTLPKELQDEQISpecifications
| ARAP2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant ARAP2. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| Q8WZ64 | |
| ARAP2 | |
| Recombinant protein corresponding to amino acids of human ARAP2. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:500-1:1000) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| ARAP2 | |
| CENTD1/FLJ13675/FLJ44916/KIAA0580/PARX | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 116984 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only