Learn More
AQP6, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant AQP6.
Brand: Abnova PAB21937.100uL
Description
The protein encoded by this gene is an aquaporin protein, which functions as a water channel in cells. Aquaporins are a family of small integral membrane proteins related to the major intrinsic protein (MIP or AQP0). This protein is specific for the kidney. This gene and related family members AQP0, AQP2, and AQP5 reside in a cluster on chromosome 12q13. [provided by RefSeq
Sequence: PDTKTLAQRLAILTGTVEVGTGAGAGAEPLKKESQPGSGAVEMESpecifications
| AQP6 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant AQP6. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| Q13520 | |
| AQP6 | |
| Recombinant protein corresponding to amino acids of human AQP6. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| AQP6 | |
| AQP2L/KID | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 363 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only