Learn More
AQP5 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of AQP5.
Brand: Abnova PAB12317.100ug
Description
Aquaporin 5 (AQP5) is a water channel protein. Aquaporins are a family of small integral membrane proteins related to the major intrinsic protein (MIP or AQP0). Aquaporin 5 plays a role in the generation of saliva, tears and pulmonary secretions. AQP0, AQP2, AQP5, and AQP6 are closely related and all map to 12q13. [provided by RefSeq
Specifications
| AQP5 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of AQP5. | |
| Lyophilized from 0.9mg NaCl, 0.2mg Na2HPO4 (5mg BSA, 0.05mg sodium azide, 0.05mg Thimerosal) | |
| AQP5 | |
| A synthetic peptide corresponding to amino acids at C-terminus of human AQP5. | |
| 100 μg | |
| Primary | |
| Different to the related rat sequence by three amino acids. | |
| Store at -20°C on dry atmosphere. After reconstitution with 200μL of deionized water and concentration will be 500μ/mL store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user. | |
| AQP5 | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 362 | |
| Human, Mouse, Rat | |
| Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only