Learn More
AQP5 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of AQP5.
Brand: Abnova PAB25157.100uL
Description
Aquaporin 5 (AQP5) is a water channel protein. Aquaporins are a family of small integral membrane proteins related to the major intrinsic protein (MIP or AQP0). Aquaporin 5 plays a role in the generation of saliva, tears and pulmonary secretions. AQP0, AQP2, AQP5, and AQP6 are closely related and all map to 12q13. [provided by RefSeq
Specifications
| AQP5 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| AQP5 | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 362 | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of AQP5. | |
| In PBS, pH 7.2 (0.05% sodium azide) | |
| AQP5 | |
| A synthetic peptide corresponding to AQP5. | |
| 100 μL | |
| Primary | |
| AQP5 polyclonal antibody detects endogenous levels of AQP5 protein. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only