missing translation for 'onlineSavingsMsg'
Learn More

APOC3 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against recombinant APOC3.

Brand:  Abnova PAB17098.100ug

Product Code. 16172290

  • £535.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Apolipoprotein C-III is a very low density lipoprotein (VLDL) protein. APOC3 inhibits lipoprotein lipase and hepatic lipase; it is thought to delay catabolism of triglyceride-rich particles. The APOA1, APOC3 and APOA4 genes are closely linked in both rat and human genomes. The A-I and A-IV genes are transcribed from the same strand, while the A-1 and C-III genes are convergently transcribed. An increase in apoC-III levels induces the development of hypertriglyceridemia. [provided by RefSeq

Specifications

Specifications

APOC3
Polyclonal
Rabbit polyclonal antibody raised against recombinant APOC3.
In 75mM PBS, 75mM NaCl, pH 7.2 (0.02% sodium azide)
APOCIII/MGC150353
Rabbit
100 μg
Primary
This antibody specifically binds to human APOC3.
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
ELISA, Western Blot
Unconjugated
Western Blot (1:2500-1:20000) ELISA (1:2500-1:20000) The optimal working dilution should be determined by the end user.
APOC3
APOC3
Recombinant protein corresponding to human APOC3.
RUO
345
Human
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only