missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APIP Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant APIP.
Brand: Abnova PAB21940.100uL
This item is not returnable.
View return policy
Description
APIP is an APAF1 (MIM 602233)-interacting protein that acts as a negative regulator of ischemic/hypoxic injury (Cho et al., 2004 [PubMed 15262985]).[supplied by OMIM
Sequence: MSGCDAREGDCCSRRCGAQDKEHPRYLIPELCKQFYHLGWVTGTGGGISLKHGDEIYIAPSGVQKERIQPEDMFVCSpecifications
| APIP | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant APIP. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| Q96GX9 | |
| APIP | |
| Recombinant protein corresponding to amino acids of human APIP. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:200-1:500) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| APIP | |
| APIP2/CGI-29/CGI29/MMRP19/dJ179L10.2 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 51074 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction