Learn More
APBA3 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant APBA3.
Brand: Abnova PAB24427.100uL
Description
The protein encoded by this gene is a member of the X11 protein family. It is an adapter protein that interacts with the Alzheimer's disease amyloid precursor protein. This gene product is believed to be involved in signal transduction processes. This gene is a candidate gene for Alzheimer's disease. [provided by RefSeq]
Sequence: QFEALPGDLVGPSPGGAPCPLHIATGHGLASQEIADAHGLLSAEAGRDDLLGLLHCEECPPSQTGPEEPLEPAPRLLQPSpecifications
| APBA3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant APBA3. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| O96018 | |
| APBA3 | |
| Recombinant protein corresponding to amino acids of human APBA3. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:200-1:500) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| APBA3 | |
| MGC:15815/X11L2/mint3 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 9546 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only