Learn More
AP1G1 Rabbit anti-Human, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of AP1G1.
Brand: Abnova PAB14334.100uL
Description
Adaptins are important components of clathrin-coated vesicles transporting ligand-receptor complexes from the plasma membrane or from the trans-Golgi network to lysosomes. The adaptin family of proteins is composed of four classes of molecules named alpha, beta-, beta prime- and gamma- adaptins. Adaptins, together with medium and small subunits, form a heterotetrameric complex called an adaptor, whose role is to promote the formation of clathrin-coated pits and vesicles. The protein encoded by this gene is a gamma-adaptin protein and it belongs to the adaptor complexes large subunits family. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: IRLRELIRTIRTARTQAEERSpecifications
| AP1G1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of AP1G1. | |
| Lyophilized from PBS | |
| O43747 | |
| AP1G1 | |
| A synthetic peptide (conjugated with diphtheria toxoid) corresponding to amino acids 5-24 of human AP1G1. | |
| RUO | |
| 164 | |
| Human, Rat | |
| Lyophilized |
| Immunofluorescence, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Immunofluorescence (1:200-1:2000) Western Blot (1:1000-1:4000) The optimal working dilution should be determined by the end user. | |
| AP1G1 | |
| ADTG/CLAPG1/MGC18255 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| This antibody has been shown to be specific to AP-1. | |
| Store at 4°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only