Learn More
ANXA5 Rabbit anti-Human, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ANXA5.
Brand: Abnova PAB7950.100ug
Description
The protein encoded by this gene belongs to the annexin family of calcium-dependent phospholipid binding proteins some of which have been implicated in membrane-related events along exocytotic and endocytotic pathways. Annexin 5 is a phospholipase A2 and protein kinase C inhibitory protein with calcium channel activity and a potential role in cellular signal transduction, inflammation, growth and differentiation. Annexin 5 has also been described as placental anticoagulant protein I, vascular anticoagulant-alpha, endonexin II, lipocortin V, placental protein 4 and anchorin CII. The gene spans 29kb containing 13 exons, and encodes a single transcript of approximately 1.6kb and a protein product with a molecular weight of about 35kDa. [provided by RefSeq
Specifications
| ANXA5 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of ANXA5. | |
| Lyophilized from 0.9mg NaCl, 0.2mg Na2HPO4 (5mg BSA, 0.05mg sodium azide, 0.05mg Thimerosal) | |
| ANX5/ENX2/PP4 | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 308 | |
| Human, Rat | |
| Lyophilized |
| Immunocytochemistry, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (0.5 ug/mL) Immunocytochemistry (0.5 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5 ug/mL) The optimal working dilution should be determined by the end user. | |
| ANXA5 | |
| ANXA5 | |
| A synthetic peptide corresponding to amino acids at middle region of human ANXA5. | |
| 100 μg | |
| Primary | |
| The synthetic peptide identical to the related rat and mouse sequence. | |
| Store at -20°C on dry atmosphere. After reconstitution with 200μL of deionized water and concentration will be 500μ/mL store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only