missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANKRD27 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ANKRD27.
Brand: Abnova PAB25707.100ug
This item is not returnable.
View return policy
Specifications
| ANKRD27 | |
| Polyclonal | |
| Unconjugated | |
| Immunohistochemistry (5 ug/mL) The optimal working dilution should be determined by the end user. | |
| ANKRD27 | |
| DKFZp434L0718/FLJ00040/PP12899/VARP | |
| Rabbit | |
| Peptide affinity purification | |
| RUO | |
| 84079 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| ELISA, Immunohistochemistry | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of ANKRD27. | |
| In PBS (0.05% sodium azide) | |
| NP_115515 | |
| ANKRD27 | |
| A synthetic peptide corresponding to 15 amino acids at internal region of human ANKRD27. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction