Learn More
ANGPT2 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ANGPT2.
Brand: Abnova PAB25796.100ug
Description
The protein encoded by this gene is an antagonist of angiopoietin 1 (ANGPT1) and endothelial TEK tyrosine kinase (TIE-2, TEK). The encoded protein disrupts the vascular remodeling ability of ANGPT1 and may induce endothelial cell apoptosis. Three transcript variants encoding three different isoforms have been found for this gene. [provided by RefSeq
Specifications
| ANGPT2 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1-2 ug/mL) Immunohistochemistry (2.5 ug/mL) The optimal working dilution should be determined by the end user. | |
| ANGPT2 | |
| AGPT2/ANG2 | |
| Rabbit | |
| Peptide affinity purification | |
| RUO | |
| 285 | |
| Human, Mouse, Rat | |
| Liquid |
| ELISA, Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of ANGPT2. | |
| In PBS (0.02% sodium azide) | |
| P_001138 | |
| ANGPT2 | |
| A synthetic peptide corresponding to 18 amino acids at C-terminus of human ANGPT2. | |
| 100 μg | |
| Primary | |
| At least three isoforms of ANGPT2 are known to exist; this antibody will detect all three isoforms. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only