Learn More
ANGPT2 Rabbit anti-Human, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ANGPT2.
Brand: Abnova PAB12280.100ug
Description
The protein encoded by this gene is an antagonist of angiopoietin 1 (ANGPT1) and endothelial TEK tyrosine kinase (TIE-2, TEK). The encoded protein disrupts the vascular remodeling ability of ANGPT1 and may induce endothelial cell apoptosis. Three transcript variants encoding three different isoforms have been found for this gene. [provided by RefSeq
Specifications
| ANGPT2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of ANGPT2. | |
| Lyophilized from 0.9mg NaCl, 0.2mg Na2HPO4 (5mg BSA, 0.05mg sodium azide, 0.05mg Thimerosal) | |
| AB009865 | |
| ANGPT2 | |
| A synthetic peptide corresponding to amino acids at C-terminus of human ANGPT2. | |
| 100 μg | |
| Primary | |
| Different from the related mouse sequence by two amino acids. | |
| Store at -20°C on dry atmosphere. After reconstitution with 200μL of deionized water and concentration will be 500μ/mL store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| Western Blot (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| ANGPT2 | |
| AGPT2/ANG2 | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 285 | |
| Human, Rat | |
| Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only