missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Angpt1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Angpt1.
Brand: Abnova PAB10092.200uL
This item is not returnable.
View return policy
Specifications
| Angpt1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of Angpt1. | |
| In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
| NP_033770 | |
| Angpt1 | |
| A synthetic peptide (conjugated with KLH) corresponding to N-terminus of mouse Angpt1. | |
| RUO | |
| 11600 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Western Blot (1:500-1:2000) Immunohistochemistry (1:200-1:800) The optimal working dilution should be determined by the end user. | |
| Angpt1 | |
| 1110046O21Rik/Ang-1/Ang1 | |
| Rabbit | |
| 200 μL | |
| Primary | |
| Human, Mouse | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction