Learn More
ANG, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ANG.
Brand: Abnova PAB25645.100ug
Description
The protein encoded by this gene is an exceedingly potent mediator of new blood vessel formation. It hydrolyzes cellular tRNAs resulting in decreased protein synthesis and is similar to pancreatic ribonuclease. Alternative splicing results in two transcript variants encoding the same protein. This gene and the gene that encodes ribonuclease, RNase A family, 4 share promoters and 5' exons. Each gene splices to a unique downstream exon that contains its complete coding region. [provided by RefSeq]
Specifications
| ANG | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| ANG | |
| ALS9/HEL168/MGC22466/MGC71966/RNASE4/RNASE5 | |
| Rabbit | |
| Peptide affinity purification | |
| RUO | |
| 283 | |
| Human, Mouse, Rat | |
| Liquid |
| ELISA, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of ANG. | |
| In PBS (0.05% sodium azide) | |
| NP_001136 | |
| ANG | |
| A synthetic peptide corresponding to 15 amino acids at internal region of human ANG. | |
| 100 μg | |
| Primary | |
| Two alternatively spliced transcript variants have been observed. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only