Learn More
AMPD1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant AMPD1.
Brand: Abnova PAB21974.100uL
Description
Adenosine monophosphate deaminase 1 catalyzes the deamination of AMP to IMP in skeletal muscle and plays an important role in the purine nucleotide cycle. Two other genes have been identified, AMPD2 and AMPD3, for the liver- and erythocyte-specific isoforms, respectively. Deficiency of the muscle-specific enzyme is apparently a common cause of exercise-induced myopathy and probably the most common cause of metabolic myopathy in the human. [provided by RefSeq
Sequence: SETSSTKLSHIDEYISSSPTYQTVPDFQRVQITGDYASGVTVEDFEIVCKGLYRALCIREKYMQKSFQRFPKTPSKYLRNIDGEAWVANESpecifications
| AMPD1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant AMPD1. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| P23109 | |
| AMPD1 | |
| Recombinant protein corresponding to amino acids of human AMPD1. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| AMPD1 | |
| MAD/MADA | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 270 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only