Learn More
AMN, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant AMN.
Brand: Abnova PAB20000.100uL
Description
The protein encoded by this gene is a type I transmembrane protein. It is thought to modulate bone morphogenetic protein (BMP) receptor function by serving as an accessory or coreceptor, and thus facilitates or hinders BMP binding. It is known that the mouse AMN gene is expressed in the extraembryonic visceral endoderm layer during gastrulation, but it is found to be mutated in amnionless mouse. The encoded protein has sequence similarity to short gastrulation (Sog) and procollagen IIA proteins in Drosophila. [provided by RefSeq
Sequence: VSKLWVPNTDFDVAANWSQNRTPCAGGAVEFPADKMVSVLVQEGHAVSDMLLPLDGELVLASGAGFGVSDVGSHLDCGAGEPAVFRDSDRFSWHDPHLWRSGDEAPGLFFVDAERVPCRHDDVFFPPSASFRVGLGPGASPVRVRSISALSpecifications
| AMN | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant AMN. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| Q9BXJ7 | |
| AMN | |
| Recombinant protein corresponding to amino acids of human AMN. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:200-1:500) The optimal working dilution should be determined by the end user. | |
| AMN | |
| PRO1028 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 81693 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only