Learn More
ALDH3B1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ALDH3B1.
Brand: Abnova PAB18080.100ug
Description
The aldehyde dehydrogenases are a family of isozymes that may play a major role in the detoxification of aldehydes generated by alcohol metabolism and lipid peroxidation. This particular gene spans about 20kb of genomic DNA and is composed of 9 coding exons. The gene is highly expressed in kidney and lung. The functional significance of this gene as well as the cellular localization of its product are presently unknown. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Specifications
| ALDH3B1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of ALDH3B1. | |
| In PBS, 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
| P43353 | |
| ALDH3B1 | |
| A synthetic peptide corresponding to human ALDH3B1. | |
| RUO | |
| 221 | |
| Human | |
| Liquid |
| ELISA, Immunohistochemistry, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (1:500∼1:1000) Immunohistochemistry (1:50∼1:100) ELISA(1:40000) The optimal working dilution should be determined by the end user. | |
| ALDH3B1 | |
| ALDH4/ALDH7/FLJ26433 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| This antibody is specific to ALDH3B1. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
For Research Use Only