Learn More
AKT2 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of AKT2.
Brand: Abnova PAB9933.100ug
Description
This gene is a putative oncogene encoding a protein belonging to a subfamily of serine/threonine kinases containing SH2-like (Src homology 2-like) domains. The gene was shown to be amplified and overexpressed in 2 of 8 ovarian carcinoma cell lines and 2 of 15 primary ovarian tumors. Overexpression contributes to the malignant phenotype of a subset of human ductal pancreatic cancers. The encoded protein is a general protein kinase capable of phophorylating several known proteins. [provided by RefSeq
Sequence: RYDSLGLLELDQRTSpecifications
| AKT2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of AKT2. | |
| In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
| PKBB/PKBBETA/PRKBB/RAC-BETA | |
| Rabbit | |
| 100 μg | |
| Primary | |
| This antibody detects both unphosphorylated and phosphorylated forms of the protein. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:4000-1:16000) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| AKT2 | |
| AKT2 | |
| A synthetic peptide (conjugated with KLH) corresponding to amino acids 455-468 of human AKT2. | |
| RUO | |
| 208 | |
| Human, Mouse, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only