missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKT1S1 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of AKT1S1.
Brand: Abnova PAB17288.100ug
This item is not returnable.
View return policy
Description
AKT1S1 is a proline-rich substrate of AKT (MIM 164730) that binds 14-3-3 protein (see YWHAH, MIM 113508) when phosphorylated (Kovacina et al., 2003 [PubMed 12524439]).[supplied by OMIM
Specifications
| AKT1S1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of AKT1S1. | |
| In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol) | |
| Q96B36 | |
| AKT1S1 | |
| A synthetic peptide corresponding to residues surrounding serine 363 of human AKT1S1. | |
| RUO | |
| 84335 | |
| Human, Mouse, Rat | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:100) ELISA (1:5000) The optimal working dilution should be determined by the end user. | |
| AKT1S1 | |
| Lobe/MGC2865/PRAS40 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| This antibody detects endogenous levels of total AKT1S1 protein. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction