Learn More
AKT1 (phospho T34), Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of AKT1.
Brand: Abnova PAB7912.100uL
Description
The serine-threonine protein kinase encoded by the AKT1 gene is catalytically inactive in serum-starved primary and immortalized fibroblasts. AKT1 and the related AKT2 are activated by platelet-derived growth factor. The activation is rapid and specific, and it is abrogated by mutations in the pleckstrin homology domain of AKT1. It was shown that the activation occurs through phosphatidylinositol 3-kinase. In the developing nervous system AKT is a critical mediator of growth factor-induced neuronal survival. Survival factors can suppress apoptosis in a transcription-independent manner by activating the serine/threonine kinase AKT1, which then phosphorylates and inactivates components of the apoptotic machinery. Multiple alternatively spliced transcript variants have been found for this gene. [provided by RefSeq
Specifications
| AKT1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of AKT1. | |
| In PBS (50% glycerol, 1mg/mL BSA, 0.05% sodium azide) | |
| AKT/MGC99656/PKB/PKB-ALPHA/PRKBA/RAC/RAC-ALPHA | |
| Rabbit | |
| 100 μL | |
| Primary | |
| This sequence is highly conserved in rat and mouse Akt. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:2000) Western Blot (1:500) The optimal working dilution should be determined by the end user. | |
| AKT1 | |
| AKT1 | |
| Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding T34 of human AKT1. | |
| RUO | |
| 207 | |
| Human, Rabbit, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only