missing translation for 'onlineSavingsMsg'
Learn More

AKT1 (phospho T308), Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic phosphopeptide of AKT1.

Brand:  Abnova PAB12601.100ug

Product Code. 16089966

  • £349.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The serine-threonine protein kinase encoded by the AKT1 gene is catalytically inactive in serum-starved primary and immortalized fibroblasts. AKT1 and the related AKT2 are activated by platelet-derived growth factor. The activation is rapid and specific, and it is abrogated by mutations in the pleckstrin homology domain of AKT1. It was shown that the activation occurs through phosphatidylinositol 3-kinase. In the developing nervous system AKT is a critical mediator of growth factor-induced neuronal survival. Survival factors can suppress apoptosis in a transcription-independent manner by activating the serine/threonine kinase AKT1, which then phosphorylates and inactivates components of the apoptotic machinery. Multiple alternatively spliced transcript variants have been found for this gene. [provided by RefSeq

Sequence: GATMKTFCG
Specifications

Specifications

AKT1
Polyclonal
Rabbit polyclonal antibody raised against synthetic phosphopeptide of AKT1.
In TBS, pH 7.2 (BSA, 10% Proclin300)
AKT/MGC99656/PKB/PKB-ALPHA/PRKBA/RAC/RAC-ALPHA
Rabbit
100 μg
Primary
The motif corresponding to the major autophosphorylation site of the human AKT1. This antibody does not cross react to the non-phosphorylated AKT1 or with other unrelated phosphorylated serine. Thr308 corresponds to GATMK in human, rat, mouse, chicken and canis of AKT/PKB proteins.
Store at 4°C for short term storage. For long term storage store at -20°C or lower.
Aliquot to avoid repeated freezing and thawing.
Dot Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Unconjugated
Western Blot (0.1-0.2 ug/mL) ELISA (0.01-0.1 ug/mL) Immunoprecipitation (2-5 ug/mL) Immunohistochemistry (1-2 ug/mL) The optimal working dilution should be determined by the end user.
AKT1
AKT1
Synthetic phosphopeptide corresponding to residues surrounding T308 of human AKT1.
RUO
207
Human, Mouse, Rat
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only