missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKT1/AKT2/AKT3 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of AKT1/AKT2/AKT3.
Brand: Abnova PAB26831.100ug
This item is not returnable.
View return policy
Description
Sequence: P-E-Yp-L-ASpecifications
| AKT1/AKT2/AKT3 | |
| Polyclonal | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:100) The optimal working dilution should be determined by the end user. | |
| P31749 (Gene ID : 207);P31751 (Gene ID : 208);Q9Y243 (Gene ID : 1000) | |
| AKT1 | |
| A synthetic peptide corresponding to residues surrounding Y315/Y316/Y312 of human AKT1/AKT2/AKT3. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of AKT1/AKT2/AKT3. | |
| In PBS, 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
| AKT/MGC99656/PKB/PKB-ALPHA/PRKBA/RAC/RAC-ALPHA | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 207/208/10000 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction