missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKT1/AKT2/AKT3 (phospho Y315/Y316/Y312), Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of AKT1/AKT2/AKT3.
Brand: Abnova PAB7764.100uL
This item is not returnable.
View return policy
Specifications
| AKT1/AKT2/AKT3 | |
| Polyclonal | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:100) The optimal working dilution should be determined by the end user. | |
| P31749 P31751 Q9Y243 | |
| AKT1 | |
| Synthetic phosphopeptide corresponding to residues surrounding Y315/Y316/Y312 of human AKT1/AKT2/AKT3. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of AKT1/AKT2/AKT3. | |
| In PBS (without mg2+ and Ca2+), 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
| AKT/MGC99656/PKB/PKB-ALPHA/PRKBA/RAC/RAC-ALPHA | |
| Rabbit | |
| Immunoaffinity purification | |
| RUO | |
| 207/208/10000 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction