Learn More
AKAP14, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of AKAP14.
Brand: Abnova PAB17405.100ug
Description
The A-kinase anchor proteins (AKAPs) are a group of structurally diverse proteins, which have the common function of binding to the regulatory subunit of protein kinase A (PKA) and confining the holoenzyme to discrete locations within the cell. This gene encodes a member of the AKAP family. The protein anchors PKA in ciliary axonemes and, in this way, may play a role in regulating ciliary beat frequency. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq
Specifications
| AKAP14 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of AKAP14. | |
| In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol) | |
| Q86UN6 | |
| AKAP14 | |
| A synthetic peptide corresponding to internal of human AKAP14. | |
| RUO | |
| 158798 | |
| Human | |
| Liquid |
| Immunofluorescence, Western Blot | |
| Unconjugated | |
| Western Blot (1:500-1:1000) Immunofluorescence (1:500-1:1000) ELISA (1:5000) The optimal working dilution should be determined by the end user. | |
| AKAP14 | |
| AKAP28 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| This antibody detects endogenous levels of total AKAP14 protein. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only