Learn More
AK1 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of AK1.
Brand:  Abnova PAB18069.100ug
Product Code. 16162600
  • £349.00 / 100µg
Description
Adenylate kinase is an enzyme involved in regulating the adenine nucleotide composition within a cell by catalyzing the reversible transfer of phosphate group among adinine nucleotides. Three isozymes of adenylate kinase have been identified in vertebrates, adenylate isozyme 1 (AK1), 2 (AK2) and 3 (AK3). AK1 is found in the cytosol of skeletal muscle, brain and erythrocytes, whereas AK2 and AK3 are found in the mitochondria of other tissues including liver and heart. AK1 was identified because of its association with a rare genetic disorder causing nonspherocytic hemolytic anemia where a mutation in the AK1 gene was found to reduce the catalytic activity of the enzyme. [provided by RefSeq
Specifications
Specifications
AK1
Polyclonal
Rabbit polyclonal antibody raised against synthetic peptide of AK1.
In PBS, 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide)
P00568
Rabbit
100 μg
Primary
This antibody is specific to AK1.
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
ELISA, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (PFA fixed)
Unconjugated
Immunohistochemistry (1:50∼1:100) Immunofluorescence (1:500∼1:1000) ELISA (1:5000) The optimal working dilution should be determined by the end user.
AK1
AK1
A synthetic peptide corresponding to human AK1.
RUO
203
Human, Mouse, Rat