Learn More
AIRE Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of AIRE.
Brand: Abnova PAB27146.100uL
Description
This gene encodes a transcriptional regulator that forms nuclear bodies and interacts with the transcriptional coactivator CBP. At least three splice variant mRNAs products have been described including one which results in a premature stop codon and a transcript predicted to be a candidate for nuclear-mediated decay (NMD). Defects in this gene cause the rare autosomal-recessive systemic autoimmune disease termed autoimmune polyendocrinopathy-candidiasis-ectodermal dystrophy (APECED). [provided by RefSeq
Specifications
| AIRE | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1:500-1:1000) Immunofluorescence (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| AIRE | |
| AIRE1/APECED/APS1/APSI/PGA1 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 326 | |
| Human, Mouse | |
| Liquid |
| Immunofluorescence, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of AIRE. | |
| In PBS, pH 7.2 (0.05% sodium azide) | |
| O43918 | |
| AIRE | |
| A synthetic peptide corresponding to human AIRE. | |
| 100 μL | |
| Primary | |
| AIRE polyclonal antibody detects endogenous levels of AIRE protein. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only