Learn More
AGTR2 Rabbit anti-Bovine, Dog, Hamster, Horse, Human, Monkey, Mouse, Pig, Rat, Sheep, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of AGTR2.
Brand: Abnova PAB25552.50ug
Description
Angiotensin II is a potent pressor hormone and a primary regulator of aldosterone secretion. It is an important effector controlling blood pressure and volume in the cardiovascular system. It acts through at least two types of receptors termed AT1 and AT2. AGTR2 belongs to a family 1 of G-protein coupled receptors. It is an integral membrane protein. It plays a role in the central nervous system and cardiovascular functions that are mediated by the renin-angiotensin system. This receptor mediates programmed cell death (apoptosis). In adults, it is highly expressed in myometrium with lower levels in adrenal gland and fallopian tube. It is highly expressed in fetal kidney and intestine. The human AGTR2 gene is composed of three exons and spans at least 5kb. Exons 1 and 2 encode for 5' untranslated mRNA sequence and exon 3 harbors the entire uninterrupted open reading frame. [provided by RefSeq]
Specifications
| AGTR2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of AGTR2. | |
| In PBS (0.09% sodium azide) | |
| AT2/ATGR2/MRX88 | |
| Rabbit | |
| Immunoaffinity chromatography | |
| RUO | |
| 186 | |
| Bovine, Canine, Equine, Hamster, Human, Monkey, Mouse, Pig, Rat, Sheep | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) The optimal working dilution should be determined by the end user. | |
| AGTR2 | |
| AGTR2 | |
| A synthetic peptide corresponding to 16 amino acids at internal region of human AGTR2. | |
| 50 μg | |
| Primary | |
| BLAST analysis of the peptide immunogen showed no homology with other human proteins, except GPR65 (63%), NPFFR2 (44%). | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only