Learn More
AES Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of AES.
Brand: Abnova PAB13166.100ug
Description
The protein encoded by this gene is similar in sequence to the amino terminus of Drosophila enhancer of split groucho, a protein involved in neurogenesis during embryonic development. The encoded protein, which belongs to the groucho/TLE family of proteins, can function as a homooligomer or as a heteroologimer with other family members to dominantly repress the expression of other family member genes. Three transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Specifications
| AES | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of AES. | |
| In PBS (0.02% sodium azide) | |
| NP_945320 | |
| AES | |
| A synthetic peptide corresponding to N-terminus 16 amino acids of human AES. | |
| RUO | |
| 166 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunocytochemistry, Western Blot | |
| Unconjugated | |
| Western Blot (2-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| AES | |
| AES-1/AES-2/ESP1/GRG/GRG5/TLE5 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only