Learn More
ADRB3 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ADRB3.
Brand: Abnova PAB25518.50ug
Description
The protein encoded by this gene belongs to the family of beta adrenergic receptors, which mediate catecholamine-induced activation of adenylate cyclase through the action of G proteins. This receptor is located mainly in the adipose tissue and is involved in the regulation of lipolysis and thermogenesis. [provided by RefSeq
Specifications
| ADRB3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of ADRB3. | |
| In PBS (0.09% sodium azide) | |
| BETA3AR | |
| Rabbit | |
| Immunoaffinity chromatography | |
| RUO | |
| 155 | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (36 ug/mL) The optimal working dilution should be determined by the end user. | |
| ADRB3 | |
| ADRB3 | |
| A synthetic peptide corresponding to 18 amino acids at C-terminus of human ADRB3. | |
| 50 μg | |
| Primary | |
| BLAST analysis of the peptide immunogen showed no homology with other human proteins, except OR2G3 (44%). | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only