Learn More
ADCY6, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ADCY6.
Brand: Abnova PAB19741.100ug
Description
This gene encodes adenylate cyclase 6, which is a membrane-associated enzyme and catalyzes the formation of the secondary messenger cyclic adenosine monophosphate (cAMP). The expression of this gene is found in normal thyroid and brain tissues, as well as some tumors; and its expression is significantly higher in one hyperfunctioning thyroid tumor than in normal thyroid tissue. Alternative splicing generates 2 transcript variants. [provided by RefSeq
Specifications
| ADCY6 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of ADCY6. | |
| In serum (0.02% sodium azide) | |
| O43306 | |
| ADCY6 | |
| A synthetic peptide corresponding to 16 amino acids at N-terminus of human ADCY6. | |
| RUO | |
| 112 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:160000) Western Blot (1:200-500) The optimal working dilution should be determined by the end user. | |
| ADCY6 | |
| AC6/DKFZp779F075/KIAA0422 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only