Learn More
ADCY10 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant ADCY10.
Brand: Abnova PAB20803.100uL
Description
The protein encoded by this gene belongs to a distinct class of mammalian adenylyl cyclase that is soluble and insensitive to G protein or forskolin regulation. It is localized in the cytoplasm and is thought to function as a general bicarbonate sensor throughout the body. It may also play an important role in the generation of cAMP in spermatozoa, implying possible roles in sperm maturation through the epididymis, capacitation, hypermotility, and/or the acrosome reaction. [provided by RefSeq
Sequence: NLENLVAQNTTGPVFCPRLYHLMAYVCILMGDGQKCGLFLNTALRLSETQGNILEKCWLNMNKESWYSTSELKEDQWLQTILSLPSWSpecifications
| ADCY10 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant ADCY10. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| Q96PN6 | |
| ADCY10 | |
| Recombinant protein corresponding to amino acids of human ADCY10. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| ADCY10 | |
| HCA2/RP1-313L4.2/SAC/SACI/Sacy | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 55811 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only