Learn More
ADAT1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant ADAT1.
Brand: Abnova PAB23711.100uL
Description
This gene is a member of the ADAR (adenosine deaminase acting on RNA) family. Using site-specific adenosine modification, proteins encoded by these genes participate in the pre-mRNA editing of nuclear transcripts. The protein encoded by this gene, tRNA-specific adenosine deaminase 1, is responsible for the deamination of adenosine 37 to inosine in eukaryotic tRNA. [provided by RefSeq
Sequence: YLLHQLQLAATLKEDSIFVPGTQKGVWKLRRDLIFVFFSSHTPCGDASIIPMLEFEDQPCCPVFRNWAHNSSVEASSNLEAPGNERKCEDPDSpecifications
| ADAT1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant ADAT1. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| Q9BUB4 | |
| ADAT1 | |
| Recombinant protein corresponding to amino acids of human ADAT1. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:20-1:50) The optimal working dilution should be determined by the end user. | |
| ADAT1 | |
| HADAT1 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 23536 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only