Learn More
ADAMTSL1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ADAMTSL1.
Brand: Abnova PAB18821.100ug
Description
This gene encodes a secreted protein and member of the ADAMTS (a disintegrin and metalloproteinase with thrombospondin motif) family. This protein lacks the metalloproteinase and disintegrin-like domains, which are typical of the ADAMTS family, but contains other ADAMTS domains, including the thrombospondin type 1 motif. This protein may have important functions in the extracellular matrix. Alternative splicing results in multiple transcript variants encoding distinct proteins. [provided by RefSeq
Specifications
| ADAMTSL1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of ADAMTSL1. | |
| In buffer containing 0.02% sodium azide | |
| ADAMTSR1/C9orf94/DKFZp686L03130/FLJ35283/FLJ41032/FLJ46891/MGC118803/MGC118805/MGC40193/PUNCTIN | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 92949 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:160000) Western Blot (1:1000-1:2000) The optimal working dilution should be determined by the end user. | |
| ADAMTSL1 | |
| ADAMTSL1 | |
| A synthetic peptide corresponding to amino acids at C-terminus of human ADAMTSL1. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only