Learn More
ADAMTS5 Rabbit anti-Human, Monkey, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ADAMTS5.
Brand: Abnova PAB25976.50ug
Description
This gene encodes a member of the ADAMTS (a disintegrin and metalloproteinase with thrombospondin motifs) protein family. Members of the family share several distinct protein modules, including a propeptide region, a metalloproteinase domain, a disintegrin-like domain, and a thrombospondin type 1 (TS) motif. Individual members of this family differ in the number of C-terminal TS motifs, and some have unique C-terminal domains. The enzyme encoded by this gene contains two C-terminal TS motifs and functions as aggrecanase to cleave aggrecan, a major proteoglycan of cartilage. [provided by RefSeq
Specifications
| ADAMTS5 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of ADAMTS5. | |
| In PBS (0.09% sodium azide) | |
| Q9UNA0 | |
| ADAMTS5 | |
| A synthetic peptide corresponding to 16 amino acids at internal region of human ADAMTS5. | |
| 50 μg | |
| Primary | |
| BLAST analysis of the peptide immunogen showed no homology with other human proteins. | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (20 ug/mL) The optimal working dilution should be determined by the end user. | |
| ADAMTS5 | |
| ADAMTS11/ADMP-2/FLJ36738 | |
| Rabbit | |
| Immunoaffinity chromatography | |
| RUO | |
| 11096 | |
| Human, Monkey | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only