Learn More
ACVR1 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ACVR1.
Brand: Abnova PAB16549.100ug
Description
Activins are dimeric growth and differentiation factors which belong to the transforming growth factor-beta (TGF-beta) superfamily of structurally related signaling proteins. Activins signal through a heteromeric complex of receptor serine kinases which include at least two type I ( I and IB) and two type II (II and IIB) receptors. These receptors are all transmembrane proteins, composed of a ligand-binding extracellular domain with cysteine-rich region, a transmembrane domain, and a cytoplasmic domain with predicted serine/threonine specificity. Type I receptors are essential for signaling; and type II receptors are required for binding ligands and for expression of type I receptors. Type I and II receptors form a stable complex after ligand binding, resulting in phosphorylation of type I receptors by type II receptors. This gene encodes activin A type I receptor which signals a particular transcriptional response in concert with activin type II receptors. Mutations in this gene are associated with fibrodysplasia ossificans progressive. [provided by RefSeq
Specifications
| ACVR1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of ACVR1. | |
| In PBS (0.02% sodium azide) | |
| NP_001096 | |
| ACVR1 | |
| A synthetic peptide corresponding to N-terminus 15 amino acids of human ACVR1. | |
| RUO | |
| 90 | |
| Human, Mouse, Rat | |
| Liquid |
| ELISA, Western Blot | |
| Unconjugated | |
| Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user. | |
| ACVR1 | |
| ACTRI/ACVR1A/ACVRLK2/ALK2/FOP/SKR1/TSRI | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Specific of ACVR1C antibody. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only