Learn More
ACTR1B, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ACTR1B.
Brand: Abnova PAB19738.100ug
Description
This gene encodes a 42.3kDa subunit of dynactin, a macromolecular complex consisting of 10 subunits ranging in size from 22 to 150kDa. Dynactin binds to both microtubules and cytoplasmic dynein and is involved in a diverse array of cellular functions, including ER-to-Golgi transport, the centripetal movement of lysosomes and endosomes, spindle formation, chromosome movement, nuclear positioning, and axonogenesis. This subunit, like ACTR1A, is an actin-related protein. These two proteins, which are of equal length and share 90% amino acid identity, are present in a constant ratio of approximately 1:15 in the dynactin complex. [provided by RefSeq
Specifications
| ACTR1B | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of ACTR1B. | |
| In serum (0.02% sodium azide) | |
| P42025 | |
| ACTR1B | |
| A synthetic peptide corresponding to C-terminus of human ACTR1B. | |
| RUO | |
| 10120 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:160000) Western Blot (1:1000-2000) The optimal working dilution should be determined by the end user. | |
| ACTR1B | |
| ARP1B/CTRN2/PC3 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only