Learn More
ACSL1 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ACSL1.
Brand: Abnova PAB25681.100ug
Description
The protein encoded by this gene is an isozyme of the long-chain fatty-acid-coenzyme A ligase family. Although differing in substrate specificity, subcellular localization, and tissue distribution, all isozymes of this family convert free long-chain fatty acids into fatty acyl-CoA esters, and thereby play a key role in lipid biosynthesis and fatty acid degradation. [provided by RefSeq
Specifications
| ACSL1 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| ACSL1 | |
| ACS1/FACL1/FACL2/LACS/LACS1/LACS2 | |
| Rabbit | |
| Peptide affinity purification | |
| RUO | |
| 2180 | |
| Human, Mouse, Rat | |
| Liquid |
| ELISA, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of ACSL1. | |
| In PBS (0.05% sodium azide) | |
| NP_001986 | |
| ACSL1 | |
| A synthetic peptide corresponding to 18 amino acids at internal region of human ACSL1. | |
| 100 μg | |
| Primary | |
| At least three isoforms of ACSL1 are known to exist; this antibody will detect all three isoforms. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only