missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACOT9, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against partial recombinant ACOT9.
Brand: Abnova PAB18533.100ug
This item is not returnable.
View return policy
Specifications
| ACOT9 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against partial recombinant ACOT9. | |
| In buffer containing 0.02% sodium azide | |
| BC012573 | |
| ACOT9 | |
| Recombinant protein corresponding to amino acids 3-212 of human ACOT9. | |
| 100 μg | |
| Primary | |
| This antibody is specific to ACOT9. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (1:500-1:1000) Immunohistochemistry (1:100-1:500) ELISA (1:20000-1:80000) The optimal working dilution should be determined by the end user. | |
| ACOT9 | |
| ACATE2/CGI-16/MT-ACT48 | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 23597 | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction