Learn More
ACCN5, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant ACCN5.
Brand: Abnova PAB27468.100uL
Description
This gene belongs to the amiloride-sensitive Na+ channel and degenerin (NaC/DEG) family, members of which have been identified in many animal species ranging from the nematode to human. The amiloride-sensitive Na(+) channel encoded by this gene is primarily expressed in the small intestine, however, its exact function is not known. [provided by RefSeq]
Sequence: AENGLLEKIKLCLSKKPLPSPTERKKFDHDFAISTSFHGIHNIVQNRSKISpecifications
| ACCN5 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant ACCN5. | |
| In PBS, pH 7.2, (40% glycerol, 0.02% sodium azide) | |
| HINAC/INAC | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 51802 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:200-1:500) The optimal working dilution should be determined by the end user. | |
| ACCN5 | |
| ACCN5 | |
| Recombinant protein corresponding to amino acids of human ACCN5. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only