Learn More
ABL1 (phospho Y412), Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of ABL1.
Brand: Abnova PAB16947.100ug
Description
The ABL1 protooncogene encodes a cytoplasmic and nuclear protein tyrosine kinase that has been implicated in processes of cell differentiation, cell division, cell adhesion, and stress response. Activity of c-Abl protein is negatively regulated by its SH3 domain, and deletion of the SH3 domain turns ABL1 into an oncogene. The t(9;22) translocation results in the head-to-tail fusion of the BCR (MIM:151410) and ABL1 genes present in many cases of chronic myelogeneous leukemia. The DNA-binding activity of the ubiquitously expressed ABL1 tyrosine kinase is regulated by CDC2-mediated phosphorylation, suggesting a cell cycle function for ABL1. The ABL1 gene is expressed as either a 6- or 7kb mRNA transcript, with alternatively spliced first exons spliced to the common exons 2-11. [provided by RefSeq
Specifications
| ABL1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of ABL1. | |
| In 20mM PBS, 0.15M NaCl, pH 7.2 (0.01% sodium azide) | |
| P00519 | |
| ABL1 | |
| Synthetic phosphopeptide corresponding to residues surrounding Y412 of human ABL1. | |
| RUO | |
| 25 | |
| Human, Mouse, Rat | |
| Liquid |
| ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:100) ELISA (1:4000) The optimal working dilution should be determined by the end user. | |
| ABL1 | |
| ABL/JTK7/bcr/abl/c-ABL/p150/v-abl | |
| Rabbit | |
| 100 μg | |
| Primary | |
| This antibody detects endogenous levels of ABL1 only when phosphorylated at tyrosine 412. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only